missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL3/CCL4 Antibody, R&D Systems™

Description
CCL3/CCL4 Monoclonal specifically detects CCL3/CCL4 in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
Specifications
| Antigen | CCL3/CCL4 |
| Applications | Western Blot, Neutralization, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 93321 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.075-0.375 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P10147 |
| Gene Symbols | CCL3 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?