missing translation for 'onlineSavingsMsg'
Learn More

Human CCL2/JE/MCP-1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18754418
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18754418 25 μg 25µg
18733139 100 μg 100µg
18766958 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18754418 Supplier R&D Systems Supplier No. MAB679SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 26 publications

CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL2/JE/MCP-1
Applications Western Blot, Immunohistochemistry, Neutralization, ELISA
Classification Monoclonal
Clone 23007
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.5-2.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF
Gene Symbols CCL2
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL2/JE/MCP-1
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6347
Test Specificity Detects human CCL2/JE/MCP-1 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant mouse (rm) CCL2, 3, 4, rhCCL3, 4, 5, 7, or 8.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.