missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CCL19 (MIP-3 beta) Recombinant Protein
Click to view available options
Quantity:
10 μg
100 μg
25 μg
500 μg
Unit Size:
100µg
10µg
25µg
500µg
Description
CCL19, also known as MIP-3 beta (Macrophage Inflammatory Protein 3 beta), is a member of the CC- subfamily of chemokines. It is most closely related to CCL21, with which it shares 32% amino acid sequence homology. CCL19 and CCL21 are expressed mainly by stromal cells in the T cell-rich zones of lymph nodes. They are critical mediators of the homeostatic trafficking of naive T cells and activated dendritic cells into the secondary lymphoid organs. CCL19 and CCL21 also play a role in T cell priming and activation, as well as the recruitment of lymphocytes to inflamed tissue. Both proteins signal via the G protein-coupled receptor, CCR7, which is expressed on T cells and mature dendritic cells. Although CCL19 and CCL21 both exhibit the same affinity for CCR7, only the binding of CCL19 results in the desensitization and internalization of the receptor.
Specifications
Specifications
| Accession Number | Q99731 |
| Concentration | 200 μg/mL |
| For Use With (Application) | Control, ELISA, Western Blot |
| Formulation | PBS with no preservative |
| Gene ID (Entrez) | 6363 |
| Molecular Weight (g/mol) | 8.8 kD |
| Name | Human MIP-3 beta (aa 22-98) |
| Quantity | 10 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction