missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CCL11 (Eotaxin) Recombinant Protein
E. coli Recombinant Protein
Brand: Invitrogen™ RP87821
Additional Details : Weight : 0.01000kg
Description
This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.Specifications
P51671 | |
Control, ELISA, Western Blot | |
6356 | |
Human Eotaxin (aa 24-97) | |
E. coli | |
RUO | |
C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11 | |
Human | |
Recombinant | |
>95% |
200 μg/mL | |
PBS with no preservative | |
8.4 kD | |
100 μg | |
-20°C | |
0.01 ng/ μg cytokine | |
CCL11 | |
Unconjugated | |
Liquid |