missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL1/I-309/TCA-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB272-100
This item is not returnable.
View return policy
Description
CCL1/I-309/TCA-3 Monoclonal specifically detects CCL1/I-309/TCA-3 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CCL1/I-309/TCA-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 1, chemokine (C-C motif) ligand 1, I-309, inflammatory cytokine I-309, P500, SCYA1, SCYA1Small-inducible cytokine A1, SCYA2, SISe, small inducible cytokine A1 (I-309, homologous to mouse Tca-3), T lymphocyte-secreted protein I-309, TCA3, TCA-3 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6346 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, ELISA | |
| 35305 | |
| Western Blot 1 ug/mL, Neutralization 0.1-0.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P22362 | |
| CCL1 | |
| E. coli-derived recombinant human CCL1/I-309/TCA-3 Lys24-Lys96 Accession # P22362 | |
| 100 μg | |
| Primary | |
| Detects human CCL1/I-309/TCA-3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human CCL2, 3, 4, 5, 7, 8, 9/10/MIP-1 gamma , 11, 14, 17, 19, 20, 21, 25, 28, recombinant mouse CCL2, 3, 5, 6, 7, 9/10/MIP-1 gamma , 11, 21, or 25 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction