missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CCDC2 Partial ORF (NP_079379, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080173-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MEEVMNGYNMLKAQNDRETQSLDVIFTERQAKEKQIRSVEEEIEQEKQATDDIIKNMSLENQVKYLEMKTTNEKLLQELDTLQQQLDSQNMKKESLEAEISpecifications
NP_079379 | |
Liquid | |
80173 | |
CCDC2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCDC2/CMG-1/CMG1/FLJ22621/MGC111562 | |
IFT74 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEEVMNGYNMLKAQNDRETQSLDVIFTERQAKEKQIRSVEEEIEQEKQATDDIIKNMSLENQVKYLEMKTTNEKLLQELDTLQQQLDSQNMKKESLEAEI | |
RUO | |
IFT74 | |
Wheat Germ (in vitro) | |
GST |