Learn More
Abnova™ Human CASP2 Partial ORF (AAH02427, 121 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000835-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms. [provided by RefSeq]
Sequence: TLSGLQHVLPPLSCDYDLSLPFPVCESCPLYKKLRLSTDTVEHSLDNKDGPLCLQVKPCTPEFYQTHFQLAYRLQSRPRGLALVLSNVHFTGEKELEFRSSpecifications
AAH02427 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TLSGLQHVLPPLSCDYDLSLPFPVCESCPLYKKLRLSTDTVEHSLDNKDGPLCLQVKPCTPEFYQTHFQLAYRLQSRPRGLALVLSNVHFTGEKELEFRS | |
RUO | |
CASP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
835 | |
CASP2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CASP-2/ICH-1L/ICH-1L/1S/ICH1/NEDD2 | |
CASP2 | |
Recombinant | |
wheat germ expression system |