Learn More
Abnova™ Human CART1 Full-length ORF (AAH10923.1, 1 a.a. - 326 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008092-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The specific function of this gene has yet to be determined in humans; however, in rodents, it is necessary for survival of the forebrain mesenchyme and may also be involved in development of the cervix. Mutations in the mouse gene lead to neural tube defects such as acrania and meroanencephaly. [provided by RefSeq]
Sequence: MEFLSEKFALKSPPSKNSDFYMGAGGPLEHVMETLDNESFYSKASAGKCVQAFGPLPRAEHHVRLERTSPCQDSSVNYGITKVEGQPLHTELNRAMDNCNSLRMSPVKGMQEKGELDELGDKCDSNVSSSKKRRHRTTFTSLQLEELEKVFQKTHYPDVYVREQLALRTELTEARVQVWFQNRRAKWRKRERYGQIQQAKSHFAATYDISVLPRTDSYPQIQNNLWAGNASGGSVVTSCMLPRDTSSCMTPYSHSPRTDSSYTGFSNHQNQFSHVPLNNFFTDSLLTGATNGHAFETKPEFERRSSSIAVLRMKAKEHTANISWAMSpecifications
AAH10923.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ALX1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8092 | |
CART1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CART1 | |
ALX1 | |
Recombinant | |
wheat germ expression system |