missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CARD14 Full-length ORF (NP_438170.1, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079092-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16144093

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein shares a similar domain structure with CARD11 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10. Two alternatively spliced variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]

Sequence: MVSSCELELQEQSLRTASDQESGDEELNRLKEENEKLRSLTFSLAEKDILEQSLDEARGSRQELVERIHSLRERAVAAERQREQYWEEKEQTLLQFQKSKMACQLYREKVNALQAQVCELQKERDQAYSARDSAQREISQSLVEKDSLRRQVFELTDQVCELRTQLRQLQAEPPGVLKQEARTREPCPREKQRLVRMHAICPRDDSDCSLVSSTESQLLSDLSATSSRELVDSFRSSSPAPPSQQSLYKRVAEDFGEEPWSFSSCLEIPEGDPGALPGAKAGDPHLDYELLDTADLPQLESSLQPVSPGRLDVSESGVLMRRRPARRILSQVTMLAFQGDALLEQISVIGGNLTGIFIHRVTPGSAADQMALRPGTQIVMVSRARPLLSPGLLMGTVAAGGVTQADFTSPRRCRSTLGWASALSWADVKRSAHL
Specifications

Specifications

NP_438170.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
74.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BIMP2/CARMA2
CARD14
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
79092
CARD14 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVSSCELELQEQSLRTASDQESGDEELNRLKEENEKLRSLTFSLAEKDILEQSLDEARGSRQELVERIHSLRERAVAAERQREQYWEEKEQTLLQFQKSKMACQLYREKVNALQAQVCELQKERDQAYSARDSAQREISQSLVEKDSLRRQVFELTDQVCELRTQLRQLQAEPPGVLKQEARTREPCPREKQRLVRMHAICPRDDSDCSLVSSTESQLLSDLSATSSRELVDSFRSSSPAPPSQQSLYKRVAEDFGEEPWSFSSCLEIPEGDPGALPGAKAGDPHLDYELLDTADLPQLESSLQPVSPGRLDVSESGVLMRRRPARRILSQVTMLAFQGDALLEQISVIGGNLTGIFIHRVTPGSAADQMALRPGTQIVMVSRARPLLSPGLLMGTVAAGGVTQADFTSPRRCRSTLGWASALSWADVKRSAHL
RUO
CARD14
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers