Learn More
Abnova™ Human CAPNS1 Partial ORF (AAH00592, 172 a.a. - 260 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000826-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Calpains are a ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. Calpain families have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. Calpain I and II are heterodimeric with distinct large subunits associated with common small subunits, all of which are encoded by different genes. This gene encodes a small subunit common to both calpain I and II and is associated with myotonic dystrophy. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq]
Sequence: KRWQAIYKQFDTDRSGTICSSELPGAFEAAGFHLNEHLYNMIIRRYSDESGNMDFDNFISCLVRLDAMFRAFKSLDKDGTGQIQVNIQESpecifications
AAH00592 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KRWQAIYKQFDTDRSGTICSSELPGAFEAAGFHLNEHLYNMIIRRYSDESGNMDFDNFISCLVRLDAMFRAFKSLDKDGTGQIQVNIQE | |
RUO | |
CAPNS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
826 | |
CAPNS1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
30K/CALPAIN4/CANP/CANPS/CAPN4/CDPS | |
CAPNS1 | |
Recombinant | |
wheat germ expression system |