Learn More
Abnova™ Human CAPN13 Partial ORF (NP_653176.2, 570 a.a. - 668 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00092291-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Calpains are a family of cytosolic calcium-activated cysteine proteases involved in a variety of cellular processes including apoptosis, cell division, modulation of integrin-cytoskeletal interactions, and synaptic plasticity (Dear et al., 2000 [PubMed 10964513]). CAPN13 belongs to the calpain large subunit family.[supplied by OMIM]
Sequence: LVHYQHVFQKVQTSPGVLLSSDLWKAIENTDFLRGIFISRELLHLVTLRYSDSVGRVSFPSLVCFLMRLEAMAKTFRNLSKDGKGLYLTEMEWMSLVMYSpecifications
NP_653176.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LVHYQHVFQKVQTSPGVLLSSDLWKAIENTDFLRGIFISRELLHLVTLRYSDSVGRVSFPSLVCFLMRLEAMAKTFRNLSKDGKGLYLTEMEWMSLVMY | |
RUO | |
CAPN13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
92291 | |
CAPN13 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23523 | |
CAPN13 | |
Recombinant | |
wheat germ expression system |