missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CADPS Partial ORF (NP_003707, 1256 a.a. - 1353 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008618-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16188895

  • 4115.00 NOK / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a novel neural/endocrine-specific cytosolic and peripheral membrane protein required for the Ca2+-regulated exocytosis of secretory vesicles. The protein acts at a stage in exocytosis that follows ATP-dependent priming, which involves the essential synthesis of phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2). Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described. [provided by RefSeq]

Sequence: RLFDQWYNSSMNVICTWLTDRMDLQLHIYQLKTLIRMVKKTYRDFRLQGVLDSTLNSKTYETIRNRLTVEEATASVSEGGGLQGISMKDSDEEDEEDD
Specifications

Specifications

NP_003707
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RLFDQWYNSSMNVICTWLTDRMDLQLHIYQLKTLIRMVKKTYRDFRLQGVLDSTLNSKTYETIRNRLTVEEATASVSEGGGLQGISMKDSDEEDEEDD
RUO
CADPS
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8618
CADPS (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CADPS1/CAPS/CAPS1/KIAA1121
CADPS
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers