missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CADM1 Partial ORF (NP_055148, 151 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023705-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: IQRDTAVEGEEIEVNCTAMASKPATTIRWFKGNTELKGKSEVEEWSDMYTVTSQLMLKVHKEDDGVPVICQVEHPAVTGNLQTQRYLEVQYKPQVHIQMTSpecifications
NP_055148 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IQRDTAVEGEEIEVNCTAMASKPATTIRWFKGNTELKGKSEVEEWSDMYTVTSQLMLKVHKEDDGVPVICQVEHPAVTGNLQTQRYLEVQYKPQVHIQMT | |
RUO | |
CADM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23705 | |
CADM1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BL2/DKFZp686F1789/IGSF4/IGSF4A/MGC149785/MGC51880/NECL2/Necl-2/RA175/ST17/SYNCAM/TSLC1/sTSLC-1/sgIGSF/synCAM1 | |
CADM1 | |
Recombinant | |
wheat germ expression system |