missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C2orf57 Full-length ORF (NP_689827.1, 1 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_689827.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 165100 |
Molecular Weight (g/mol) | 68kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16149013
|
Abnova™
H00165100-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16139013
|
Abnova™
H00165100-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: MALPGYPLGNVDDSRSKDSPAGEPQGQVPLTADVLAVSSSVASTDWQDIDQASFKTATPRAISTSGDKDKSAVVPEHGQKTPRKITPLLPSQNPSPLQVSMSLQNPAWDRQVQDARTSQSLVVFPSHLLGKDKMSQMASVPEREPESAPSAPSAELQSTQHMEAQPVESDADHVTAGANGQHGPQAASTTKSAEEKAEHPKAPHPEAEALPSDESPVAMGANVVDSLGDLQTWFFPPPPAGSVSPSPGPHEVALGRRPLDPSLYTASEENSYMRSMTSLLDRGEGSISSLADILVWSETTMGMAIATGFLDSGHSTVADLLHSSGPSLRSVPSLVGSVSSAFSSGLVSGTSSALRTITRVLETVEQRTVEGIRSAMRYLTSHLTPRQAQADPNYDSpecifications
NP_689827.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68kDa | |
Glutathione Sepharose 4 Fast Flow | |
MALPGYPLGNVDDSRSKDSPAGEPQGQVPLTADVLAVSSSVASTDWQDIDQASFKTATPRAISTSGDKDKSAVVPEHGQKTPRKITPLLPSQNPSPLQVSMSLQNPAWDRQVQDARTSQSLVVFPSHLLGKDKMSQMASVPEREPESAPSAPSAELQSTQHMEAQPVESDADHVTAGANGQHGPQAASTTKSAEEKAEHPKAPHPEAEALPSDESPVAMGANVVDSLGDLQTWFFPPPPAGSVSPSPGPHEVALGRRPLDPSLYTASEENSYMRSMTSLLDRGEGSISSLADILVWSETTMGMAIATGFLDSGHSTVADLLHSSGPSLRSVPSLVGSVSSAFSSGLVSGTSSALRTITRVLETVEQRTVEGIRSAMRYLTSHLTPRQAQADPNYD | |
RUO | |
C2orf57 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
165100 | |
C2orf57 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC35154 | |
C2orf57 | |
Recombinant | |
wheat germ expression system |