Learn More
Abnova™ Human C20orf42 Partial ORF (NP_060141.2, 180 a.a. - 269 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand:  Abnova™ H00055612-Q01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16128236
  • 6245.00 NOK / 25µg
Estimated Shipment: 26-06-2024
Description
Sequence: PGLYSKTMTPIYDPINGTPASSTMTWFSDSPLTEQNCSILAFSQPPQSPEALADMYQPRSLVDKAKLNAGWLDSSRSLMEQGIQEDEQLL
Specifications
Specifications
NP_060141.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PGLYSKTMTPIYDPINGTPASSTMTWFSDSPLTEQNCSILAFSQPPQSPEALADMYQPRSLVDKAKLNAGWLDSSRSLMEQGIQEDEQLL
RUO
FERMT1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
55612
C20orf42 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C20orf42/DTGCU2/FLJ20116/FLJ23423/KIND1/UNC112A/URP1
FERMT1
Recombinant
wheat germ expression system