missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C18orf23 Partial ORF (NP_689683, 122 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00147341-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: VSHPRRSQERVSVHPHRLHPSFDFGQLQTPQPRYLAEGTDWDLSVDAGLSPAQFQVRPIPQHYQHYLATPRMHHFPRNSSSTQMVVHEIRNYPYPQLHFSpecifications
NP_689683 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VSHPRRSQERVSVHPHRLHPSFDFGQLQTPQPRYLAEGTDWDLSVDAGLSPAQFQVRPIPQHYQHYLATPRMHHFPRNSSSTQMVVHEIRNYPYPQLHF | |
RUO | |
C18orf23 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
147341 | |
C18orf23 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ34218 | |
C18orf23 | |
Recombinant | |
wheat germ expression system |