missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C10orf97 Full-length ORF (NP_079224.1, 1 a.a. - 445 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080013-P01.25ug
Additional Details : Weight : 0.00010kg
Description
C10ORF97 is a member of the caspase-associated recruitment domain (CARD) family of proteins that play a role in apoptosis (Liu et al., 2002 [PubMed 12054670]).[supplied by OMIM]
Sequence: MSELTKELMELVWGTKSSPGLSDTIFCRWTQGFVFSESEGSALEQFEGGPCAVIAPVQAFLLKKLLFSSEKSSWRDCSEEEQKELLCHTLCDILESACCDHSGSYCLVSWLRGKTTEETASISGSPAESSCQVEHSSALAVEELGFERFHALIQKRSFRSLPELKDAVLDQYSMWGNKFGVLLFLYSVLLTKGIENIKNEIEDASEPLIDPVYGHGSQSLINLLLTGHAVSNVWDGDRECSGMKLLGIHEQAAVGFLTLMEALRYCKVGSYLKSPKFPIWIVGSETHLTVFFAKDMALVAPEAPSEQARRVFQTYDPEDNGFIPDSLLEDVMKALDLVSDPEYINLMKNKLDPEGLGIILLGPFLQEFFPDQGSSGPESFTVYHYNGLKQSNYNEKVMYVEGTAVVMGFEDPMLQTDDTPIKRCLQTKWPYIELLWTTDRSPSLNSpecifications
NP_079224.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CARP/DERP5/FLJ13397/MST126/MSTP126/RP11-394I23.1/my042 | |
C10orf97 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
80013 | |
C10orf97 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSELTKELMELVWGTKSSPGLSDTIFCRWTQGFVFSESEGSALEQFEGGPCAVIAPVQAFLLKKLLFSSEKSSWRDCSEEEQKELLCHTLCDILESACCDHSGSYCLVSWLRGKTTEETASISGSPAESSCQVEHSSALAVEELGFERFHALIQKRSFRSLPELKDAVLDQYSMWGNKFGVLLFLYSVLLTKGIENIKNEIEDASEPLIDPVYGHGSQSLINLLLTGHAVSNVWDGDRECSGMKLLGIHEQAAVGFLTLMEALRYCKVGSYLKSPKFPIWIVGSETHLTVFFAKDMALVAPEAPSEQARRVFQTYDPEDNGFIPDSLLEDVMKALDLVSDPEYINLMKNKLDPEGLGIILLGPFLQEFFPDQGSSGPESFTVYHYNGLKQSNYNEKVMYVEGTAVVMGFEDPMLQTDDTPIKRCLQTKWPYIELLWTTDRSPSLN | |
RUO | |
C10orf97 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |