Learn More
Abnova™ Human C10orf63 Full-length ORF (NP_659447.1, 1 a.a. - 256 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00219670-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Enkurin interacts with transient receptor potential canonical (TRPC) cation channels (see TRPC1; MIM 602343) and functions as an adaptor protein, tethering signal transduction proteins to TRPC channels (Sutton et al., 2004 [PubMed 15385169]).[supplied by OMIM]
Sequence: MDPTCSSECIYNLIPSDLKEPPQPPRYISIFKATVKDDMQKAKTAMKTMGPAKVEVPSPKDFLKKHSKEKTLPPKKNFDRNVPKKPAVPLKTDHPVMGIQSGKNFINTNAADIIMGVAKKPKPIYVDKRTGDKHDLEPSGLVPKYINKKDYGVTPEYICKRNEEIKKAQEDYDRYIQENLKKAAMKRLSDEEREAVLQGLKKNWEEVHKEFQSLSVFIDSIPKKIRKQRLEEEMKQLEHDIGIIEKHKIIYIANNASpecifications
NP_659447.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781F21103/MGC26778 | |
C10orf63 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
219670 | |
C10orf63 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDPTCSSECIYNLIPSDLKEPPQPPRYISIFKATVKDDMQKAKTAMKTMGPAKVEVPSPKDFLKKHSKEKTLPPKKNFDRNVPKKPAVPLKTDHPVMGIQSGKNFINTNAADIIMGVAKKPKPIYVDKRTGDKHDLEPSGLVPKYINKKDYGVTPEYICKRNEEIKKAQEDYDRYIQENLKKAAMKRLSDEEREAVLQGLKKNWEEVHKEFQSLSVFIDSIPKKIRKQRLEEEMKQLEHDIGIIEKHKIIYIANNA | |
RUO | |
C10orf63 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |