Learn More
Human Brachyury Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB20851-100
Product Code. 18764187
  • 4560.00 NOK / 100µg
Description
Brachyury Monoclonal specifically detects Brachyury in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Brachyury
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
brachyury protein, MGC104817, Protein T, SAVA, T brachyury (mouse) homolog, T brachyury homolog, T Brachyury Transcription Factor, T, brachyury homolog (mouse), T-Box Transcription Factor T, TBXT, TF, TFT
Rabbit
Protein A or G purified from cell culture supernatant
Primary
Detects human Brachyury in direct ELISAs.
Human
Supernatant
Immunocytochemistry
1161B
Immunocytochemistry 8-25 μg;/mL
O15178
T
E. coli-derived recombinant human Brachyury, Met230-Met435, Accession No. O15178
100 μg
6862.0
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.