Learn More
Abnova™ Human BHLHB8 Partial ORF (NP_803238.1, 75 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00168620-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
MIST1 is a basic helix-loop-helix (bHLH) protein that binds to the DNA target sequence CANNTG (E box) present in the promoter or enhancer region of numerous developmentally regulated genes.
Sequence: QRRLESNERERQRMHKLNNAFQALREVIPHVRADKKLSKIETLTLAKNYIKSLTATILTMSSSRLPSpecifications
NP_803238.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QRRLESNERERQRMHKLNNAFQALREVIPHVRADKKLSKIETLTLAKNYIKSLTATILTMSSSRLP | |
RUO | |
BHLHA15 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
168620 | |
BHLHB8 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BHLHB8/MIST1 | |
BHLHA15 | |
Recombinant | |
wheat germ expression system |