missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human BDNF Recombinant Protein

Product Code. 11588836
Change view
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11588836 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11588836 Supplier Gibco™ Supplier No. 10908010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Human BDNF Recombinant Protein

Human BDNF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

Recombinant Human Brain-Derived Neurotrophic Factor (BDNF) is a bioactive protein intended for use in cell culture applications. BDNF is a neurotrophic factor involved in the survival and differentiation of neurons in the central and peripheral nervous system and used in the maintenance of motor neurons and the differentiation of both cholinergic and dopaminergic neurons.
  • High purity—no interference from other proteins or contaminants
  • High biological activity—more results with less protein
  • Proven compatibility—Gibco™ proteins bioassayed with Gibco™ media
High Purity Recombinant Bioactive Protein
Recombinant Human Brain-Derived Neurotrophic Factor is one of our more than 250 available Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.

High Biological Activity
In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Brain-Derived Neurotrophic Factor (BDNF) specific activity is determined by measuring the dose-dependent induction of choline acetyl transferase activity in primary septal cell cultures.

Proven Compatibility
Gibco™ proteins are bioassayed with Gibco™ media. Since 1962, Gibco™ has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Purification Method Membrane-Filtered
Quantity 5 μg
Gene Alias Brain-Derived Neurotrophic Factor (BDNF)
Species Human
Content And Storage Store in freezer (-5 to -30°C).
Classification Carrier-Free
Endotoxin Level < 0.1 ng/μg
Mass 5 μg
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Expression System E. coli
Protein Family Growth Factors & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Neurotrophic Factors
Research Category Neurobiology, Stem Cell Research, Differentiation
Purity or Quality Grade 96 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.