Learn More
Abnova™ Human BCS1L Full-length ORF (NP_004319.1, 1 a.a. - 419 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000617-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a homolog of the S. cerevisiae bcs1 protein which is involved in the assembly of complex III of the mitochondrial respiratory chain. The encoded protein does not contain a mitochondrial targeting sequence but experimental studies confirm that it is imported into mitochondria. Mutations in this gene are associated with mitochondrial complex III deficiency and the GRACILE syndrome. Two alternatively spliced transcripts encoding the same protein have been described. [provided by RefSeq]
Sequence: MPLSDFILALKDNPYFGAGFGLVGVGTALALARKGVQLGLVAFRRHYMITLEVPARDRSYAWLLSWLTRHSTRTQHLSVETSYLQHESGRISTKFEFVPSPGNHFIWYRGKWIRVERSREMQMIDLQTGTPWESVTFTALGTDRKVFFNILEEARELALQQEEGKTVMYTAVGSEWRPFGYPRRRRPLNSVVLQQGLADRIVRDVQEFIDNPKWYTDRGIPYRRGYLLYGPPGCGKSSFITALAGELEHSICLLSLTDSSLSDDRLNHLLSVAPQQSLVLLEDVDAAFLSRDLAVENPVKYQGLGRLTFSGLLNALDGVASTEARIVFMTTNHVDRLDPALIRPGRVDLKEYVGYCSHWQLTQMFQRFYPGQAPSLAENFAEHVLRATNQISPAQVQGYFMLYKNDPVGAIHNAESLRRSpecifications
NP_004319.1 | |
Liquid | |
617 | |
BCS1L (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCS/BCS1/BJS/FLNMS/GRACILE/Hs.6719/PTD/h-BCS | |
BCS1L | |
Yes | |
wheat germ expression system |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
BCS1L | |
Wheat Germ (in vitro) | |
GST |