missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BCL2L12 Full-length ORF (NP_001035758.1, 1 a.a. - 333 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_001035758.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 83596 |
Molecular Weight (g/mol) | 63.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16135393
|
Abnova™
H00083596-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16125393
|
Abnova™
H00083596-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains a Bcl-2 homology domain 2 (BH2). The function of this gene has not yet been determined. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]
Sequence: MGRPAGLFPPLCPFLGFRPEACWERHMQIERAPSVPPFLRWAGYRPGPVRRRGKVELIKFVRVQWRRPQVEWRRRRWGPGPGASMAGSEELGLREDTLRVLAAFLRRGEAAGSPVPTPPSPAQEEPTDFLSRLRRCLPCSLGRGAAPSESPRPCSLPIRPCYGLEPGPATPDFYALVAQRLEQLVQEQLKSPPSPELQGPPSTEKEAILRRLVALLEEEAEVINQKLASDPALRSKLVRLSSDSFARLVELFCSRDDSSRPSRACPGPPPPSPEPLARLALAMELSRRVAGLGGTLAGLSVEHVHSFTPWIQAHGGWEGILAVSPVDLNLPLDSpecifications
NP_001035758.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC120313/MGC120314/MGC120315 | |
BCL2L12 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
83596 | |
BCL2L12 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
BCL2L12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |