Learn More
Abnova™ Human BAIAP3 Partial ORF (NP_003924, 302 a.a. - 410 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008938-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This p53-target gene encodes a brain-specific angiogenesis inhibitor. The protein is a seven-span transmembrane protein and a member of the secretin receptor family. It interacts with the cytoplasmic region of brain-specific angiogenesis inhibitor 1. This protein also contains two C2 domains, which are often found in proteins involved in signal transduction or membrane trafficking. Its expression pattern and similarity to other proteins suggest that it may be involved in synaptic functions. [provided by RefSeq]
Sequence: LVEACRKLNEVIGLKGMGRYFKQIVKSARANGTAGPTEDHTDDFLGCLNIPVREVPVAGVDRWFKLEPRSSASRVQGHCHLVLKLITTQRDTAMSQRGRSGFLSHLLLLSpecifications
NP_003924 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LVEACRKLNEVIGLKGMGRYFKQIVKSARANGTAGPTEDHTDDFLGCLNIPVREVPVAGVDRWFKLEPRSSASRVQGHCHLVLKLITTQRDTAMSQRGRSGFLSHLLLL | |
RUO | |
BAIAP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8938 | |
BAIAP3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BAP3/KIAA0734/MGC138334 | |
BAIAP3 | |
Recombinant | |
wheat germ expression system |