Learn More
ACROBiosystems Human BAFFR / TNFRSF13C Protein, Llama IgG2b Fc Tag, low endotoxin
low endotoxin
Brand:  ACROBiosystems BAR-H5258-100ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16447209
  • 5341.70 NOK / 100µg
Estimated Shipment: 07-08-2024
to see stock.
Description
Description
BAFF receptor (B-cell activating factor receptor, BAFF-R), also known as tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), is a membrane protein of the TNF receptor superfamily which recognizes BAFF. B-cell activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. Overexpression of BAFF in mice results in mature B-cell hyperplasia and symptoms of systemic lupus erythematosus (SLE). Also, some SLE patients have increased levels of BAFF in serum. Therefore, it has been proposed that abnormally high levels of BAFF may contribute to the pathogenesis of autoimmune diseases by enhancing the survival of autoreactive B cells.
This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 34.4 kDa. The protein migrates as 42-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications
Specifications
NP_443177.1
100 μg
Ser 7 - Ala 71
BAFFR,TNFRSF13C,BROMIX,CD268,CVID4,prolixin,BAFF-R
Unconjugated
Llama IgG Fc Tag
Less than 0.01 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
34.4 kDa
Human BAFFR, Llama IgG2b Fc Tag, low endotoxin (BAR-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).
-20°C
BAFFR
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Other Recombinant Proteins