Learn More
ACROBiosystems Human BAFFR / TNFRSF13C Protein, Llama IgG2b Fc Tag, low endotoxin
low endotoxin
Brand: ACROBiosystems BAR-H5258-1mg
Additional Details : Weight : 0.00010kg
Description
BAFF receptor (B-cell activating factor receptor, BAFF-R), also known as tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), is a membrane protein of the TNF receptor superfamily which recognizes BAFF. B-cell activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. Overexpression of BAFF in mice results in mature B-cell hyperplasia and symptoms of systemic lupus erythematosus (SLE). Also, some SLE patients have increased levels of BAFF in serum. Therefore, it has been proposed that abnormally high levels of BAFF may contribute to the pathogenesis of autoimmune diseases by enhancing the survival of autoreactive B cells.
This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 34.4 kDa. The protein migrates as 42-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_443177.1 | |
1 mg | |
Ser 7 - Ala 71 | |
BAFFR,TNFRSF13C,BROMIX,CD268,CVID4,prolixin,BAFF-R | |
Unconjugated | |
Llama IgG Fc Tag | |
Less than 0.01 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
34.4 kDa | |
Human BAFFR, Llama IgG2b Fc Tag, low endotoxin (BAR-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1). | |
-20°C | |
BAFFR | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |