Learn More
Abnova™ Human Bach2 Full-Length Orf (Aai66613.1, 1 A.A.-841 A.A.) Recombinant Protein With Gst Tag At N-Terminal.
Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAI66613.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 60468 |
Name | BACH2 (Human) Recombinant Protein (P01) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16113213
|
Abnova™
H00060468-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16123213
|
Abnova™
H00060468-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: MSVDEKPDSPMYVYESTVHCTNILLGLNDQRKKDILCDVTLIVERKEFRAHRAVLAACSEYFWQALVGQTKNDLVVSLPEEVTARGFGPLLQFAYTAKLLLSRENIREVIRCAEFLRMHNLEDSCFSFLQTQLLNSEDGLFVCRKD
Specifications
AAI66613.1 | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. | |
BACH2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
RUO | |
BACH2 | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
60468 | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BACH2 | |
GST |
For Research Use Only