Learn More
Abnova™ Human B3GALT4 Partial ORF (NP_003773.1, 151 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008705-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the beta-1,3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. The carbohydrate chains synthesized by these enzymes are designated as type 1, whereas beta4GalT enzymes synthesize type 2 carbohydrate chains. The ratio of type 1:type 2 chains changes during embryogenesis. By sequence similarity, the beta3GalT genes fall into at least two groups: beta3GalT4 and 4 other beta3GalT genes (beta3GalT1-3, beta3GalT5). This gene is oriented telomere to centromere in close proximity to the ribosomal protein S18 gene. The functionality of the encoded protein is limited to ganglioseries glycolipid biosynthesis. [provided by RefSeq]
Sequence: TLKTLSGLNWAEKHCPMARYVLKTDDDVYVNVPELVSELVLRGGRWGQWERSTEPQREAEQEGGQVLHSEEVPLLYLGRVHWRVNPSRTPGGRHRVSEEQSpecifications
NP_003773.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TLKTLSGLNWAEKHCPMARYVLKTDDDVYVNVPELVSELVLRGGRWGQWERSTEPQREAEQEGGQVLHSEEVPLLYLGRVHWRVNPSRTPGGRHRVSEEQ | |
RUO | |
B3GALT4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8705 | |
B3GALT4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DJ1033B10.3/Gal-T2/GalT4/beta3GALT4/beta3Gal-T4 | |
B3GALT4 | |
Recombinant | |
wheat germ expression system |