missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AZIN1 Partial ORF (NP_056962, 339 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051582-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16127146

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ornithine decarboxylase (ODC) catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. Ornithine decarboxylase antizymes play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. The protein encoded by this gene is highly similar to ODC. It binds to ODC antizyme and stabilizes ODC, thus inhibiting antizyme-mediated ODC degradation. Two alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]

Sequence: HKKYKEDEPLFTSSLWGPSCDELDQIVESCLLPELNVGDWLIFDNMGADSFHEPSAFNDFQRPAIYYMMSFSDWYEMQDAGITSDSMMKNFFFVPSCIQLSQEDSFSAE
Specifications

Specifications

NP_056962
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HKKYKEDEPLFTSSLWGPSCDELDQIVESCLLPELNVGDWLIFDNMGADSFHEPSAFNDFQRPAIYYMMSFSDWYEMQDAGITSDSMMKNFFFVPSCIQLSQEDSFSAE
RUO
AZIN1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
51582
AZIN1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC3832/MGC691/OAZI/OAZIN/ODC1L
AZIN1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers