Learn More
Abnova™ Human AXIN2 Partial ORF (NP_004646, 611 a.a. - 710 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008313-Q02.25ug
Additional Details : Weight : 0.00010kg
Description
The Axin-related protein, Axin2, presumably plays an important role in the regulation of the stability of beta-catenin in the Wnt signaling pathway, like its rodent homologs, mouse conductin/rat axil. In mouse, conductin organizes a multiprotein complex of APC (adenomatous polyposis of the colon), beta-catenin, glycogen synthase kinase 3-beta, and conductin, which leads to the degradation of beta-catenin. Apparently, the deregulation of beta-catenin is an important event in the genesis of a number of malignancies. The AXIN2 gene has been mapped to 17q23-q24, a region that shows frequent loss of heterozygosity in breast cancer, neuroblastoma, and other tumors. Mutations in this gene have been associated with colorectal cancer with defective mismatch repair. [provided by RefSeq]
Sequence: EEGDRSQDVWQWMLESERQSKPKPHSAQSTKKAYPLESARSSPGERASRHHLWGGNSGHPRTTPRAHLFTQDPAMPPLTPPNTLAQLEEACRRLAEVSKPSpecifications
NP_004646 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EEGDRSQDVWQWMLESERQSKPKPHSAQSTKKAYPLESARSSPGERASRHHLWGGNSGHPRTTPRAHLFTQDPAMPPLTPPNTLAQLEEACRRLAEVSKP | |
RUO | |
AXIN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8313 | |
AXIN2 (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AXIL/DKFZp781B0869/MGC10366/MGC126582 | |
AXIN2 | |
Recombinant | |
wheat germ expression system |