missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ATP9B Full-length ORF (AAH53561.1, 1 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00374868-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120374

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MPLMMSEEGFENEESDYHTLPRARIMQRKRGLEWFVCDGWKFLCTSCCGWLINICRRKKELKARTVWLGCPEKCEEKHPRNSIKNQKYNVFTFIPGVLYEQFKFFLNLYFLVISCSQFVPALKIGYLYTYWAPLMT
Specifications

Specifications

AAH53561.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
42.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ATP9B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
374868
ATP9B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATPASEP/ATPIIB/DKFZp686H2093/FLJ46612/HUSSY-20/MGC150650/MGC150651/MGC61572/NEO1L
ATP9B
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers