Learn More
Abnova™ Human ATP6V1F Partial ORF (NP_004222, 10 a.a. - 119 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009296-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c″, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is the V1 domain F subunit protein. [provided by RefSeq]
Sequence: VIGDEDTVTGFLLGGIGELNKNRHPNFLVVEKDTTINEIEDTFRQFLNRDDIGIILINQYIAEMVRHALDAHQQSIPAVLEIPSKEHPYDAAKDSILRRARGMFTAEDLRSpecifications
NP_004222 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VIGDEDTVTGFLLGGIGELNKNRHPNFLVVEKDTTINEIEDTFRQFLNRDDIGIILINQYIAEMVRHALDAHQQSIPAVLEIPSKEHPYDAAKDSILRRARGMFTAEDLR | |
RUO | |
ATP6V1F | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9296 | |
ATP6V1F (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP6S14/MGC117321/MGC126037/MGC126038/VATF/Vma7 | |
ATP6V1F | |
Recombinant | |
wheat germ expression system |