Learn More
Abnova™ Human ASB11 Partial ORF (NP_543149, 214 a.a. - 323 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00140456-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: KLLELGASVDHGQWLDTPLHAAARQSNVEVIHLLTDYGANLKRRNAQGKSALDLAAPKSSVEQALLLREGPPALSQLCRLCVRKCLGRACHQAIHKLHLPEPLERFLLYQSpecifications
NP_543149 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KLLELGASVDHGQWLDTPLHAAARQSNVEVIHLLTDYGANLKRRNAQGKSALDLAAPKSSVEQALLLREGPPALSQLCRLCVRKCLGRACHQAIHKLHLPEPLERFLLYQ | |
RUO | |
ASB11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
140456 | |
ASB11 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp779E2460/MGC119168/MGC119169 | |
ASB11 | |
Recombinant | |
wheat germ expression system |