missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ART4 Full-length ORF (NP_066549.2, 1 a.a. - 314 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_066549.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 420 |
Molecular Weight (g/mol) | 62.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101931
|
Abnova™
H00000420-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16191921
|
Abnova™
H00000420-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that contains a mono-ADP-ribosylation (ART) motif. It is a member of the ADP-ribosyltransferase gene family but enzymatic activity has not been demonstrated experimentally. Antigens of the Dombrock blood group system are located on the gene product, which is glycosylphosphatidylinosotol-anchored to the erythrocyte membrane. Allelic variants, some of which lead to adverse transfusion reactions, are known. [provided by RefSeq]
Sequence: MGPLINRCKKILLPTTVPPATMRIWLLGGLLPFLLLLSGLQRPTEGSEVAIKIDFDFAPGSFDDQYQGCSKQVMEKLTQGDYFTKDIEAQKNYFRMWQKAHLAWLNQGKVLPQNMTTTHAVAILFYTLNSNVHSDFTRAMASVARTPQQYERSFHFKYLHYYLTSAIQLLRKDSIMENGTLCYEVHYRTKDVHFNAYTGATIRFGQFLSTSLLKEEAQEFGNQTLFTIFTCLGAPVQYFSLKKEVLIPPYELFKVINMSYHPRGDWLQLRSTGNLSTYNCQLLKASSKKCIPDPIAIASLSFLTSVIIFSKSRVSpecifications
NP_066549.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD297/DO/DOK1 | |
ART4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
420 | |
ART4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ART4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |