missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARR3 Partial ORF (NP_004303, 178 a.a. - 259 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000407-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: GPSAQTIRRFLLSAQPLQLQAWMDREVHYHGEPISVNVSINNCTNKVIKKIKISVDQITDVVLYSLDKYTKTVFIQEFTETVSpecifications
NP_004303 | |
Liquid | |
407 | |
ARR3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARRX | |
ARR3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.76kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GPSAQTIRRFLLSAQPLQLQAWMDREVHYHGEPISVNVSINNCTNKVIKKIKISVDQITDVVLYSLDKYTKTVFIQEFTETV | |
RUO | |
ARR3 | |
Wheat Germ (in vitro) | |
GST |