missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARPM2 Partial ORF (NP_536356, 209 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_536356 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 140625 |
Molecular Weight (g/mol) | 35.75kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16141947
|
Abnova™
H00140625-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16131947
|
Abnova™
H00140625-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this intronless gene belongs to the actin family. Studies have shown that this protein may be involved in cytoskeletal organization similar to other cytoplasmic actin-related protein (ARP) subfamily members. Antibody raised against the human protein has been used to detect the protein by immunoblotting and immunofluorescence microscopy, demonstrating its specific synthesis in the testis, late in spermatid differentiation, and its localization in the calyx. [provided by RefSeq]
Sequence: DKGLVDDIKKKLCYVALEPEKELSRRPEEVLREYKLPDGNIISLGDPLHQAPEALFVPQQLGSQSPGLSNMVSSSITKCDTDIQKILFGEISpecifications
NP_536356 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARPM2/ARPT2/Arp-T2/FLJ25424/HARPM2 | |
ACTRT2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
140625 | |
ARPM2 (Human) Recombinant Protein (Q01) | |
DKGLVDDIKKKLCYVALEPEKELSRRPEEVLREYKLPDGNIISLGDPLHQAPEALFVPQQLGSQSPGLSNMVSSSITKCDTDIQKILFGEI | |
RUO | |
ACTRT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |