missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARNT2 Full-length ORF (AAH36099, 1 a.a. - 716 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009915-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134222

  • 5540.00 NOK / 25µg
Estimated Shipment: 21-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the basic-helix-loop-helix-Per-Arnt-Sim (bHLH-PAS) superfamily of transcription factors. The encoded protein acts as a partner for several sensor proteins of the bHLH-PAS family, forming heterodimers with the sensor proteins that bind regulatory DNA sequences in genes responsive to developmental and environmental stimuli. Under hypoxic conditions, the encoded protein complexes with hypoxia-inducible factor 1alpha in the nucleus and this complex binds to hypoxia-responsive elements in enhancers and promoters of oxygen-responsive genes. A highly similar protein in mouse forms functional complexes with both aryl hydrocarbon receptors and Single-minded proteins, suggesting addition roles for the encoded protein in the metabolism of xenobiotic compounds and the regulation of neurogenesis, respectively. [provided by RefSeq]

Sequence: MATPAAVNPPEMASDIPGSVTLPVAPMAATGQVRMAGAMPARGGKRRSGMDFDDEDGEGPSKFSRENHSEIERRRRNKMTQYITELSDMVPTCSALARKPDKLTILRMAVSHMKSMRGTGNKSTDGAYKPSFLTEQELKHLILEAADGFLFVVAAETGRVIYVSDSVTPVLNQPQSEWFGSTLYEQVHPDDVEKLREQLCTSENSMTGRILDLKTGTVKKEGQQSSMRMCMGSRRSFICRMRCGNAPLDHLPLNRITTMRKRFRNGLGPVKEGEAQYAVVHCTGYIKAWPPAGMTIPEEDADVGQGSKYCLVAIGRLQVTSSPVCMDMNGMSVPTEFLSRHNSDGIITFVDPRCISVIGYQPQDLLGKDILEFCHPEDQSHLRESFQQVVKLKGQVLSVMYRFRTKNREWMLIRTSSFTFQNPYSDEIEYIICTNTNVKQLQQQQAELEVHQRDGLSSYDLSQVPVPNLPAGVHEAGKSVEKADAIFSQERDPRFAEMFAGISASEKKMMSSASAAGTQQIYSQGSPFPSGHSGKAFSSSVVHVPGVNDIQSSSSTGQNMSQISRQLNQSQVAWTGSRPPFPGQQIPSQSSKTQSSPFGIGTSHTYPADPSSYSPLSSPATSSPSGNAYSSLANRTPGFAESGQSSGQFQGRPSEVWSQWQSQHHGQQSGEQHSHQQPGQTEVFQDMLPMPGDPGNEWWSPHSRQHFRQPISYARM
Specifications

Specifications

AAH36099
Liquid
9915
ARNT2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0307/bHLHe1
ARNT2
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
104.28kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ARNT2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers