Learn More
Abnova™ Human ARL4C Full-length ORF (NP_005728.2, 1 a.a. - 192 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010123-P01.25ug
Additional Details : Weight : 0.00010kg
Description
ADP-ribosylation factor-like 4C is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4C is closely similar to ARL4A and ARL4D and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in cholesterol transport. [provided by RefSeq]
Sequence: MGNISSNISAFQSLHIVMLGLDSAGKTTVLYRLKFNEFVNTVPTIGFNTEKIKLSNGTAKGISCHFWDVGGQEKLRPLWKSYSRCTDGIIYVVDSVDVDRLEEAKTELHKVTKFAENQGTPLLVIANKQDLPKSLPVAEIEKQLALHELIPATTYHVQPACAIIGEGLTEGMDKLYEMILKRRKSLKQKKKRSpecifications
NP_005728.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ARL4C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10123 | |
ARL4C (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARL7/LAK | |
ARL4C | |
Recombinant | |
wheat germ expression system |