missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARID3B Full-length ORF (AAH60824.1, 1 a.a. - 253 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH60824.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10620 |
Molecular Weight (g/mol) | 54.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16195432
|
Abnova™
H00010620-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16185432
|
Abnova™
H00010620-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the ARID (AT-rich interaction domain) family of DNA-binding proteins. The encoded protein is homologous with two proteins that bind to the retinoblastoma gene product, and also with the mouse Bright and Drosophila dead ringer proteins. A pseudogene on chromosome 1p31 exists for this gene. Members of the ARID family have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification. [provided by RefSeq]
Sequence: MEPLQQQQQQQQQQQKQPHLAPLQMDAREKQGQQMREAQFLYAQKLVTQPTLLSATAGRPSGSTPLGPLARVPPTAAVAQVFERGNMNSEPEEEDGGLEDEDGDDEVAEVAEKETQAASKYFHVQKVARQDPRVAPMSNLLPAPGLPPHGQQAKEDHTKDASKASPSVSTAGQPNWNLDEQLKQNGGLAWSDDADGGRGREISRDFAKLYELDGDPERKEFLDDLFVFMQKRGECASTHHSNSGNTDRVPTVCSpecifications
AAH60824.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BDP/DRIL2 | |
ARID3B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10620 | |
ARID3B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ARID3B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |