missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARID1B Partial ORF (NP_059989, 1364 a.a. - 1460 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057492-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: PPAKRHEGDMYNMQYSSQQQEMYNQYGGSYSGPDRRPIQGQYPYPYSRERMQGPGQIQTHGIPPQMMGGPLQSSSSEGPQQNMWAARNDMPYPYQNRSpecifications
NP_059989 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PPAKRHEGDMYNMQYSSQQQEMYNQYGGSYSGPDRRPIQGQYPYPYSRERMQGPGQIQTHGIPPQMMGGPLQSSSSEGPQQNMWAARNDMPYPYQNR | |
RUO | |
ARID1B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57492 | |
ARID1B (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
6A3-5/BAF250b/BRIGHT/DAN15/ELD/OSA1/KIAA1235/p250R | |
ARID1B | |
Recombinant | |
wheat germ expression system |