Learn More
Abnova™ Human ARHGAP17 Full-length ORF (NP_001006635.1, 1 a.a. - 881 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055114-P01.25ug
Additional Details : Weight : 0.00010kg
Description
RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA (MIM 165390), RAC1 (MIM 602048), and CDC42 (MIM 116952).[supplied by OMIM]
Sequence: MKKQFNRMKQLANQTVGRAEKTEVLSEDLLQIERRLDTVRSICHHSHKRLVACFQGQHGTDAERRHKKLPLTALAQNMQEASTQLEDSLLGKMLETCGDAENQLALELSQHEVFVEKEIVDPLYGIAEVEIPNIQKQRKQLARLVLDWDSVRARWNQAHKSSGTNFQGLPSKIDTLKEEMDEAGNKVEQCKDQLAADMYNFMAKEGEYGKFFVTLLEAQADYHRKALAVLEKTLPEMRAHQDKWAEKPAFGTPLEEHLKRSGREIALPIEACVMLLLETGMKEEGLFRIGAGASKLKKLKAALDCSTSHLDEFYSDPHAVAGALKSYLRELPEPLMTFNLYEEWTQVASVQDQDKKLQDLWRTCQKLPPQNFVNFRYLIKFLAKLAQTSDVNKMTPSNIAIVLGPNLLWARNEGTLAEMAAATSVHVVAVIEPIIQHADWFFPEEVEFNVSEAFVPLTTPSSNHSFHTGNDSDSGTLERKRPASMAVMEGDLVKKESFGVKLMDFQAHRRGGTLNRKHISPAFQPPLPPTDGSTVVPAGPEPPPQSSRAESSSGGGTVPSSAGILEQGPSPGDGSPPKPKDPVSAAVPAPGRNNSQIASGQNQPQAAAGSHQLSMGQPHNAAGPSPHTLRRAVKKPAPAPPKPGNPPPGHPGGQSSSGTSQHPPSLSPKPPTRSPSPPTQHTGQPPGQPSAPSQLSAPRRYSSSLSPIQAPNHPPPQPPTQATPLMHTKPNSQGPPNPMALPSEHGLEQPSHTPPQTPTPPSTPPLGKQNPSLPAPQTLAGGNPETAQPHAGTLPRPRPVPKPRNRPSVPPPPQPPGVHSAGDSSLTNTAPTASKIVTDSNSRVSEPHRSIFPEMHSDSASKDVPGRILLDIDNDTESTALSpecifications
NP_001006635.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
121.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ARHGAP17 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55114 | |
ARHGAP17 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp564A1363/FLJ37567/FLJ43368/MGC87805/MST066/MST110/MSTP038/MSTP066/MSTP110/NADRIN/RICH1/WBP15 | |
ARHGAP17 | |
Recombinant | |
wheat germ expression system |