Learn More
Human Aquaporin 4/AQP4 Antibody, Novus Biologicals Biologicals™
Product Code. 30306105
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
Product Code. Quantity unitSize
30306105 25 μg 25µg
30306119 100 μg 100µg
2 options
Product Code. 30306105
Brand: R&D Systems MAB11659SP
Mouse Monoclonal Antibody
Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Multiplex Immunofluorescence,Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Antigen Aquaporin-4
Applications Multiplex Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 1093722
Conjugate Unconjugated
Dilution Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 25 μg/mL
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2μm filtered solution in PBS.
Gene Accession No. P55087
Gene Alias AQP4, AQP-4, aquaporin 4, MGC22454, MIWC, WCH4
Host Species Mouse
Immunogen Synthetic Peptide, Accession # P55087
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361
Reconstitution Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Product Type Antibody
Form Purified
Isotype IgG