Learn More
Abnova™ Human AQP7 Full-length ORF (NP_001161.1, 1 a.a. - 342 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000364-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Aquaporins/major intrinsic protein (MIP) are a family of water-selective membrane channels. Aquaporin 7 has greater sequence similarity with AQP3 and AQP9 and they may be a subfamily. Aquaporin 7 and AQP3 are at the same chromosomal location suggesting that 9p13 may be a site of an aquaporin cluster. Aquaporin 7 facilitates water, glycerol and urea transport. It may play an important role in sperm function. [provided by RefSeq]
Sequence: MVQASGHRRSTRGSKMVSWSVIAKIQEILQRKMVREFLAEFMSTYVMMVFGLGSVAHMVLNKKYGSYLGVNLGFGFGVTMGVHVAGRISGAHMNAAVTFANCALGRVPWRKFPVYVLGQFLGSFLAAATIYSLFYTAILHFSGGQLMVTGPVATAGIFATYLPDHMTLWRGFLNEAWLTGMLQLCLFAITDQENNPALPGTEALVIGILVVIIGVSLGMNTGYAINPSRDLPPRIFTFIAGWGKQVFSNGENWWWVPVVAPLLGAYLGGIIYLVFIGSTIPREPLKLEDSVAYEDHGITVLPKMGSHEPTISPLTPVSVSPANRSSVHPAPPLHESMALEHFSpecifications
NP_001161.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
AQP7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
364 | |
AQP7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AQP7L/AQP9/AQPap/MGC149555/MGC149556 | |
AQP7 | |
Recombinant | |
wheat germ expression system |