Learn More
Abnova™ Human APOL3 Full-length ORF (NP_055164.1, 1 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080833-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the apolipoprotein L gene family, and it is present in a cluster with other family members on chromosome 6. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids, including cholesterol, and/or allow the binding of lipids to organelles. In addition, expression of this gene is up-regulated by tumor necrosis factor-alpha in endothelial cells lining the normal and atherosclerotic iliac artery and aorta. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MDSEKKRFTEEATKYFRERVSPVHLQILLTNNEAWKRFVTAAELPRDEADALYEALKKLRTYAAIEDEYVQQKDEQFREWFLKEFPQVKRKIQESIEKLRALANGIEEVHRGCTISNVVSSSTGAASGIMSLAGLVLAPFTAGTSLALTAAGVGLGAASAVTGITTSIVEHSYTSSAEAEASRLTATSIDRLKVFKEVMRDITPNLLSLLNNYYEATQTIGSEIRAIRQARARARLPVTTWRISAGSGGQAERTIAGTTRAVSRGARILSATTSGIFLALDVVNLVYESKHLHEGAKSASAEELRRQAQELEENLMELTQIYQRLNPCHTHSpecifications
NP_055164.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
APOL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
80833 | |
APOL3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APOLIII/CG12-1 | |
APOL3 | |
Recombinant | |
wheat germ expression system |