Learn More
Abnova™ Human AP1GBP1 Partial ORF (NP_009178.3, 456 a.a. - 555 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011276-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that interacts with the gamma subunit of AP1 clathrin-adaptor complex. The AP1 complex is located at the trans-Golgi network and associates specific proteins with clathrin-coated vesicles. This encoded protein may act to connect the AP1 complex to other proteins. Alternatively spliced transcript variants that encode different isoforms have been described for this gene. [provided by RefSeq]
Sequence: DDFQDFQDASKSGSLDDSFSDFQELPASSKTSNSQHGNSAPSLLMPLPGTKALPSMDKYAVFKGIAADKSSENTVPPGDPGDKYSAFRELEQTAENKPLGSpecifications
NP_009178.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DDFQDFQDASKSGSLDDSFSDFQELPASSKTSNSQHGNSAPSLLMPLPGTKALPSMDKYAVFKGIAADKSSENTVPPGDPGDKYSAFRELEQTAENKPLG | |
RUO | |
AP1GBP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11276 | |
AP1GBP1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC104959/SYNG | |
AP1GBP1 | |
Recombinant | |
wheat germ expression system |