Learn More
Abnova™ Human ANAPC5 Full-length ORF (NP_057321.2, 1 a.a. - 755 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051433-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a tetratricopeptide repeat-containing component of the anaphase promoting complex/cyclosome (APC/C), a large E3 ubiquitin ligase that controls cell cycle progression by targeting a number of cell cycle regulators such as B-type cyclins for 26S proteasome-mediated degradation through ubiquitination. The encoded protein is required for the proper ubiquitination function of APC/C and for the interaction of APC/C with transcription coactivators. It also interacts with polyA binding protein and represses internal ribosome entry site-mediated translation. Multiple transcript variants encoding different isoforms have been found for this gene. These differences cause translation initiation at a downstream AUG and result in a shorter protein (isoform b), compared to isoform a. [provided by RefSeq]
Sequence: MASVHESLYFNPMMTNGVVHANVFGIKDWVTPYKIAVLVLLNEMSRTGEGAVSLMERRRLNQLLLPLLQGPDITLSKLYKLIEESCPQLANSVQIRIKLMAEGELKDMEQFFDDLSDSFSGTEPEVHKTSVVGLFLRHMILAYSKLSFSQVFKLYTALQQYFQNGEKKTVEDADMELTSRDEGERKMEKEELDVSVREEEVSCSGPLSQKQAEFFLSQQASLLKNDETKALTPASLQKELNNLLKFNPDFAEAHYLSYLNNLRVQDVFSSTHSLLHYFDRLILTGAESKSNGEEGYGRSLRYAALNLAALHCRFGHYQQAELALQEAIRIAQESNDHVCLQHCLSWLYVLGQKRSDSYVLLEHSVKKAVHFGLPYLASLGIQSLVQQRAFAGKTANKLMDALKDSDLLHWKHSLSELIDISIAQKTAIWRLYGRSTMALQQAQMLLSMNSLEAVNAGVQQNNTESFAVALCHLAELHAEQGCFAAASEVLKHLKERFPPNSQHAQLWMLCDQKIQFDRAMNDGKYHLADSLVTGITALNSIEGVYRKAVVLQAQNQMSEAHKLLQKLLVHCQKLKNTEMVISVLLSVAELYWRSSSPTIALPMLLQALALSKEYRLQYLASETVLNLAFAQLILGIPEQALSLLHMAIEPILADGAILDKGRAMFLVAKCQVASAASYDQPKKAEALEAAIENLNEAKNYFAKVDCKERIRDVVYFQARLYHTLGKTQERNRCAMLFRQLHQELPSHGVPLINHLSpecifications
NP_057321.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
111.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ANAPC5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51433 | |
ANAPC5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APC5 | |
ANAPC5 | |
Recombinant | |
wheat germ expression system |