missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AMBN Full-length ORF (NP_057603.1, 1 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_057603.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 258 |
Molecular Weight (g/mol) | 74.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16191521
|
Abnova™
H00000258-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16101531
|
Abnova™
H00000258-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ameloblastin is thought to represent an unique ameloblast-specific gene product that may be important in enamel matrix formation and mineralization. The gene is located on chromosome 4 near other genes associated with mineralized tissues: osteopontin, bone sialoprotein, and bone morphogenetic protein 3. Based on its cytogenetic location, this gene is a candidate gene for one form of the disorder, dentinogenesis imperfecta, and/or the disorder, autosomal dominant amylogenesis imperfecta. [provided by RefSeq]
Sequence: MSASKIPLFKMKDLILILCLLEMSFAVPFFPQQSGTPGMASLSLETMRQLGSLQRLNTLSQYSRYGFGKSFNSLWMHGLLPPHSSLPWMRPREHETQQYEYSLPVHPPPLPSQPSLKPQQPGLKPFLQSAAATTNQATALKEALQPPIHLGHLPLQEGELPLVQQQVAPSDKPPKPELPGVDFADPQGPSLPGMDFPDPQGPSLPGLDFADPQGSTIFQIARLISHGPMPQNKQSPLYPGMLYVPFGANQLNAPARLGIMSSEEVAGGREDPMAYGAMFPGFGGMRPGFEGMPHNPAMGGDFTLEFDSPVAATKGPENEEGGAQGSPMPEANPDNLENPAFLTELEPAPHAGLLALPKDDIPGLPRSPSGKMKGLPSVTPAAADPLMTPELADVYRTYDADMTTSVDFQEEATMDTTMAPNSLQTSMPGNKAQEPEMMHDAWHFQEPSpecifications
NP_057603.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
74.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AMBN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
258 | |
AMBN (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
AMBN | |
Recombinant | |
wheat germ expression system |