Learn More
Human alpha 2-Macroglobulin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB19381-100
 View more versions of this product
Product Code. 18710117
  • 6155.00 NOK / 100µg
Description
alpha 2-Macroglobulin Monoclonal antibody specifically detects alpha 2-Macroglobulin in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha 2-Macroglobulin
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2
Human
Purified
ELISA
257303
ELISA
A2M, alpha 2Macroglobulin, Alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5, CPAMD5DKFZp779B086, FWP007, S863-7
Human plasma-derived alpha 2-Macroglobulin
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG1