Learn More
Abnova™ Human ALDH18A1 Partial ORF (NP_002851, 696 a.a. - 795 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005832-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the aldehyde dehydrogenase family and encodes a bifunctional ATP- and NADPH-dependent mitochondrial enzyme with both gamma-glutamyl kinase and gamma-glutamyl phosphate reductase activities. The encoded protein catalyzes the reduction of glutamate to delta1-pyrroline-5-carboxylate, a critical step in the de novo biosynthesis of proline, ornithine and arginine. Mutations in this gene lead to hyperammonemia, hypoornithinemia, hypocitrullinemia, hypoargininemia and hypoprolinemia and may be associated with neurodegeneration, cataracts and connective tissue diseases. Alternatively spliced transcript variants, encoding different isoforms, have been described for this gene. [provided by RefSeq]
Sequence: TDVIVTEDENTAEFFLQHVDSACVFWNASTRFSDGYRFGLGAEVGISTSRIHARGPVGLEGLLTTKWLLRGKDHVVSDFSEHGSLKYLHENLPIPQRNTNSpecifications
NP_002851 | |
Liquid | |
5832 | |
ALDH18A1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GSAS/MGC117316/P5CS/PYCS | |
ALDH18A1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TDVIVTEDENTAEFFLQHVDSACVFWNASTRFSDGYRFGLGAEVGISTSRIHARGPVGLEGLLTTKWLLRGKDHVVSDFSEHGSLKYLHENLPIPQRNTN | |
RUO | |
ALDH18A1 | |
Wheat Germ (in vitro) | |
GST |