Learn More
Abnova™ Human ALAS1 Partial ORF (NP_000679, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000211-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Delta-aminolevulinate synthase (ALAS; EC 2.3.1.37) catalyzes the condensation of glycine with succinyl-CoA to form delta-aminolevulinic acid. This nuclear-encoded mitochondrial enzyme is the first and rate-limiting enzyme in the mammalian heme biosynthetic pathway. There are 2 tissue-specific isozymes: a housekeeping enzyme encoded by the ALAS1 gene and an erythroid tissue-specific enzyme encoded by ALAS2 (MIM 301300).[supplied by OMIM]
Sequence: MESVVRRCPFLSRVPQAFLQKAGKSLLFYAQNCPKMMEVGAKPAPRALSTAAVHYQQIKETPPASEKDKTAKAKVQQTPDGSQQSPDGTQLPSGHPLPSpecifications
NP_000679 | |
Liquid | |
211 | |
ALAS1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALAS/ALAS3/ALASH/MIG4 | |
ALAS1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MESVVRRCPFLSRVPQAFLQKAGKSLLFYAQNCPKMMEVGAKPAPRALSTAAVHYQQIKETPPASEKDKTAKAKVQQTPDGSQQSPDGTQLPSGHPLP | |
RUO | |
ALAS1 | |
Wheat Germ (in vitro) | |
GST |